Anti-NDUFV1 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-3959R-CY5
BOSSBS-3959R-CY5EA 900 EUR
BOSSBS-3959R-CY5
Anti-NDUFV1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I) that is believed to belong to the minimal assembly required for catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NDUFV1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR