Anti-NDUFV2 Rabbit Polyclonal Antibody (Cy7®)
Lieferant: Bioss
BS-3960R-CY7
BOSSBS-3960R-CY7EA 900 EUR
BOSSBS-3960R-CY7
Anti-NDUFV2 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
The NADH-ubiquinone oxidoreductase complex (complex I) of the mitochondrial respiratory chain catalyzes the transfer of electrons from NADH to ubiquinone, and consists of at least 43 subunits. The complex is located in the inner mitochondrial membrane. This gene encodes the 24 kDa subunit of complex I, and is involved in electron transfer. Mutations in this gene are implicated in Parkinson's disease, bipolar disorder, schizophrenia, and have been found in one case of early onset hypertrophic cardiomyopathy and encephalopathy. A non-transcribed pseudogene of this locus is found on chromosome 19. [provided by RefSeq, Oct 2009].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NDUFV2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat