Anti-SDHA Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-3970R-CY5
BOSSBS-3970R-CY5EA 900 EUR
BOSSBS-3970R-CY5
Anti-SDHA Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Flavoprotein (FP) subunit of succinate dehydrogenase (SDH) that is involved in complex II of the mitochondrial electron transport chain and is responsible for transferring electrons from succinate to ubiquinone (coenzyme Q). Can act as a tumor suppressor.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SDHA
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat