Anti-SDHA Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-3970R-CY5.5
BOSSBS-3970R-CY5.5EA 900 EUR
BOSSBS-3970R-CY5.5
Anti-SDHA Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Flavoprotein (FP) subunit of succinate dehydrogenase (SDH) that is involved in complex II of the mitochondrial electron transport chain and is responsible for transferring electrons from succinate to ubiquinone (coenzyme Q). Can act as a tumor suppressor.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SDHA
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR