Anti-FBP1 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-3980R-CY5
BOSSBS-3980R-CY5EA 900 EUR
BOSSBS-3980R-CY5
Anti-FBP1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Catalyzes the hydrolysis of fructose 1,6-bisphosphate to fructose 6-phosphate in the presence of divalent cations, acting as a rate-limiting enzyme in gluconeogenesis. Plays a role in regulating glucose sensing and insulin secretion of pancreatic beta-cells. Appears to modulate glycerol gluconeogenesis in liver. Important regulator of appetite and adiposity; increased expression of the protein in liver after nutrient excess increases circulating satiety hormones and reduces appetite-stimulating neuropeptides and thus seems to provide a feedback mechanism to limit weight gain.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FBP1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR