Anti-GALK1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-3983R-A488
BOSSBS-3983R-A488EA 893 EUR
BOSSBS-3983R-A488
Anti-GALK1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Galactokinase is a major enzyme for the metabolism of galactose and its deficiency causes congenital cataracts in the adult population. GALK1 sequence shares the greatest level of conservation, 44.5% identity with that from E. coli and 34.6% amino acid identity with the product of the human GALK2 gene.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Galactose kinase
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR