Anti-SLC25A10 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-3997R-A750
BOSSBS-3997R-A750EA 890 EUR
BOSSBS-3997R-A750
Anti-SLC25A10 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Involved in translocation of malonate, malate and succinate in exchange for phosphate, sulfate, sulfite or thiosulfate across mitochondrial inner membrane.

Type: Primary
Antigen: SLC25A10
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-287/287
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR