Anti-SLC25A10 Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BS-3997R-CY3
BOSSBS-3997R-CY3EA 900 EUR
BOSSBS-3997R-CY3
Anti-SLC25A10 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Involved in translocation of malonate, malate and succinate in exchange for phosphate, sulfate, sulfite or thiosulfate across mitochondrial inner membrane.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SLC25A10
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat