Anti-UGP2 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-4043R
BOSSBS-4043REA 725 EUR
BOSSBS-4043R
Anti-UGP2 Rabbit Polyclonal Antibody
Antikörper
UGP2 (UDP glucose pyrophosphorylase 2) is an important intermediary in cellular metabolic pathways. It transfers a glucose moiety from glucose-1-phosphate to MgUTP and forms UDP glucose and MgPPi.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: UGP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR