Anti-UGP2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Lieferant: Bioss
BS-4043R-A350
BOSSBS-4043R-A350EA 893 EUR
BOSSBS-4043R-A350
Anti-UGP2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
UGP2 (UDP glucose pyrophosphorylase 2) is an important intermediary in cellular metabolic pathways. It transfers a glucose moiety from glucose-1-phosphate to MgUTP and forms UDP glucose and MgPPi.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: UGP2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat