Anti-UGP2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss
BS-4043R-FITC
BOSSBS-4043R-FITCEA 900 EUR
BOSSBS-4043R-FITC
Anti-UGP2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
UGP2 (UDP glucose pyrophosphorylase 2) is an important intermediary in cellular metabolic pathways. It transfers a glucose moiety from glucose-1-phosphate to MgUTP and forms UDP glucose and MgPPi.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: UGP2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR