Anti-HACL1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-4046R-A647
BOSSBS-4046R-A647EA 893 EUR
BOSSBS-4046R-A647
Anti-HACL1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
2 Hydroxy phytanoyl CoA lyase catalyzes a carbon-carbon cleavage reaction. It cleaves a 2 hydroxy 3 methylacyl CoA into formyl CoA and a 2 methyl branched fatty aldehyde.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HPCL2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR