Anti-PPAR alpha Ser12 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-4055R-A750
BOSSBS-4055R-A750EA 917 EUR
BOSSBS-4055R-A750
Anti-PPAR alpha Ser12 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Ligand-activated transcription factor. Key regulator of lipid metabolism. Activated by the endogenous ligand 1-palmitoyl-2-oleoyl-sn-glycerol-3-phosphocholine (16:0/18:1-GPC). Activated by oleylethanolamide, a naturally occurring lipid that regulates satiety. Receptor for peroxisome proliferators such as hypolipidemic drugs and fatty acids. Regulates the peroxisomal beta-oxidation pathway of fatty acids. Functions as transcription activator for the ACOX1 and P450 genes. Transactivation activity requires heterodimerization with RXRA and is antagonized by NR2C2. May be required for the propagation of clock information to metabolic pathways regulated by PER2.

Type: Primary
Antigen: PPAR alpha Ser12
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR