Anti-ZBTB8A Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Lieferant: Bioss
BS-4057R-A488
BOSSBS-4057R-A488EA 893 EUR
BOSSBS-4057R-A488
Anti-ZBTB8A Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
DEGS1 is a member of the membrane fatty acid desaturase family which is responsible for inserting double bonds into specific positions in fatty acids. It contains three His containing consensus motifs that are characteristic of a group of membrane fatty acid desaturases. It has sphingolipid-delta-4-desaturase activity and converts D-erythro-sphinganine to D-erythro-sphingosine (E-sphing-4-enine).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ZBTB8A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat