Anti-ZBTB8A Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-4057R-A647
BOSSBS-4057R-A647EA 893 EUR
BOSSBS-4057R-A647
Anti-ZBTB8A Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
DEGS1 is a member of the membrane fatty acid desaturase family which is responsible for inserting double bonds into specific positions in fatty acids. It contains three His containing consensus motifs that are characteristic of a group of membrane fatty acid desaturases. It has sphingolipid-delta-4-desaturase activity and converts D-erythro-sphinganine to D-erythro-sphingosine (E-sphing-4-enine).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZBTB8A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR