Anti-ZBTB8A Rabbit Polyclonal Antibody (Cy5.5®)
Lieferant: Bioss
BS-4057R-CY5.5
BOSSBS-4057R-CY5.5EA 900 EUR
BOSSBS-4057R-CY5.5
Anti-ZBTB8A Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
DEGS1 is a member of the membrane fatty acid desaturase family which is responsible for inserting double bonds into specific positions in fatty acids. It contains three His containing consensus motifs that are characteristic of a group of membrane fatty acid desaturases. It has sphingolipid-delta-4-desaturase activity and converts D-erythro-sphinganine to D-erythro-sphingosine (E-sphing-4-enine).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ZBTB8A
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat