Anti-GPD2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-4061R-A680
BOSSBS-4061R-A680EA 890 EUR
BOSSBS-4061R-A680
Anti-GPD2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
The protein encoded by this gene localises to the inner mitochondrial membrane and catalyses the conversion of glycerol-3-phosphate to dihydroxyacetone phosphate, using FAD as a cofactor. Along with GDP1, the encoded protein constitutes the glycerol phosphate shuttle, which reoxidises NADH formed during glycolysis. Two transcript variants encoding the same protein have been found for this gene.
Type: Primary
Antigen: GPD2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 165-270/727
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat