Anti-GPD2 Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BS-4061R-CY3
BOSSBS-4061R-CY3EA 893 EUR
BOSSBS-4061R-CY3
Anti-GPD2 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
The protein encoded by this gene localizes to the inner mitochondrial membrane and catalyzes the conversion of glycerol-3-phosphate to dihydroxyacetone phosphate, using FAD as a cofactor. Along with GDP1, the encoded protein constitutes the glycerol phosphate shuttle, which reoxidizes NADH formed during glycolysis. Two transcript variants encoding the same protein have been found for this gene.[provided by RefSeq, Jan 2010].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GPD2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat