Anti-Glycerol kinase Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-4062R-A680
BOSSBS-4062R-A680EA 890 EUR
BOSSBS-4062R-A680
Anti-Glycerol kinase Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Glycerol kinase catalyses the formation of glycerol 3 phosphate from ATP and glycerol. Dihydroxyacetone and L glyceraldehyde can also act as acceptors; UTP and, in the case of the yeast enzyme, ITP and GTP can act as donors. It provides a way for glycerol derived from fats or glycerides to enter the glycolytic pathway.

Type: Primary
Antigen: Glycerol kinase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 21-120/559
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR