Anti-HMGCR Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-4063R-CY3
BOSSBS-4063R-CY3EA 929 EUR
BOSSBS-4063R-CY3
Anti-HMGCR Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Transmembrane glycoprotein that is the rate-limiting enzyme in cholesterol biosynthesis as well as in the biosynthesis of nonsterol isoprenoids that are essential for normal cell function including ubiquinone and geranylgeranyl proteins.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HMGCR
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR