Anti-HMGCR Rabbit Polyclonal Antibody (Cy7®)
Lieferant: Bioss
BS-4063R-CY7
BOSSBS-4063R-CY7EA 929 EUR
BOSSBS-4063R-CY7
Anti-HMGCR Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Transmembrane glycoprotein that is the rate-limiting enzyme in cholesterol biosynthesis as well as in the biosynthesis of nonsterol isoprenoids that are essential for normal cell function including ubiquinone and geranylgeranyl proteins.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HMGCR
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat