Anti-LATS1/2 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-4081R
BOSSBS-4081REA 725 EUR
BOSSBS-4081R
Anti-LATS1/2 Rabbit Polyclonal Antibody
Antikörper
This gene encodes a serine/threonine protein kinase belonging to the LATS tumor suppressor family. The protein localizes to centrosomes during interphase, and early and late metaphase. It interacts with the centrosomal proteins aurora-A and ajuba and is required for accumulation of gamma-tubulin and spindle formation at the onset of mitosis. It also interacts with a negative regulator of p53 and may function in a positive feedback loop with p53 that responds to cytoskeleton damage. Additionally, it can function as a co-repressor of androgen-responsive gene expression. [provided by RefSeq].

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: LATS1/2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR