Anti-PLPP3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-4127R-A350
BOSSBS-4127R-A350EA 893 EUR
BOSSBS-4127R-A350
Anti-PLPP3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Catalyzes the conversion of phosphatidic acid (PA) to diacylglycerol (DG). In addition it hydrolyzes lysophosphatidic acid (LPA), ceramide-1-phosphate (C-1-P) and sphingosine-1-phosphate (S-1-P). The relative catalytic efficiency is LPA = PA >C-1-P >S-1-P. May be involved in cell adhesion and in cell-cell interactions.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PLPP3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR