Anti-PLPP3 Rabbit Polyclonal Antibody (Cy5.5®)
Lieferant: Bioss
BS-4127R-CY5.5
BOSSBS-4127R-CY5.5EA 900 EUR
BOSSBS-4127R-CY5.5
Anti-PLPP3 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Catalyzes the conversion of phosphatidic acid (PA) to diacylglycerol (DG). In addition it hydrolyzes lysophosphatidic acid (LPA), ceramide-1-phosphate (C-1-P) and sphingosine-1-phosphate (S-1-P). The relative catalytic efficiency is LPA = PA >C-1-P >S-1-P. May be involved in cell adhesion and in cell-cell interactions.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PLPP3
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse