Anti-GLUL Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-4143R-A647
BOSSBS-4143R-A647EA 893 EUR
BOSSBS-4143R-A647
Anti-GLUL Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
This enzyme has 2 functions: it catalyzes the production of glutamine and 4-aminobutanoate (gamma-aminobutyric acid, GABA), the latter in a pyridoxal phosphate-independent manner (By similarity). Essential for proliferation of fetal skin fibroblasts.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Glutamine synthetase
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR