Anti-Glutamine synthetase Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-4143R-A750
BOSSBS-4143R-A750EA 890 EUR
BOSSBS-4143R-A750
Anti-Glutamine synthetase Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
This enzyme has 2 functions: it catalyses the production of glutamine and 4-aminobutanoate (gamma-aminobutyric acid, GABA), the latter in a pyridoxal phosphate-independent manner (By similarity). Essential for proliferation of foetal skin fibroblasts.

Type: Primary
Antigen: Glutamine synthetase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 285-373/373
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR