Anti-GLUL Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-4143R-CY5
BOSSBS-4143R-CY5EA 900 EUR
BOSSBS-4143R-CY5
Anti-GLUL Rabbit Polyclonal Antibody (Cy5®)
Antikörper
This enzyme has 2 functions: it catalyzes the production of glutamine and 4-aminobutanoate (gamma-aminobutyric acid, GABA), the latter in a pyridoxal phosphate-independent manner (By similarity). Essential for proliferation of fetal skin fibroblasts.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Glutamine synthetase
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat