Anti-SLC52A3 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-4164R
BOSSBS-4164REA 725 EUR
BOSSBS-4164R
Anti-SLC52A3 Rabbit Polyclonal Antibody
Antikörper
Transporter for riboflavin, which must be obtained as a nutrient via intestinal absorption. Riboflavin transport is Na(+)-independent at low pH but significantly reduced by Na(+) depletion under neutral pH conditions. Activity is strongly inhibited by riboflavin analogs, such as lumiflavin, flavin mononucleotide (FMN), flavin adenine dinucleotide (FAD), by methylene blue, and to a lesser extent by amiloride.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: SLC52A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR