Anti-P2RY2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Lieferant: Bioss
BS-4180R-A488
BOSSBS-4180R-A488EA 901 EUR
BOSSBS-4180R-A488
Anti-P2RY2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Receptor for ATP and UTP coupled to G-proteins that activate a phosphatidylinositol-calcium second messenger system. The affinity range is UTP = ATP >ATP-gamma-S >2-methylthio-ATP = ADP.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: P2RY2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat