Anti-P2RY2 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-4180R-CY5
BOSSBS-4180R-CY5EA 909 EUR
BOSSBS-4180R-CY5
Anti-P2RY2 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Receptor for ATP and UTP coupled to G-proteins that activate a phosphatidylinositol-calcium second messenger system. The affinity range is UTP = ATP >ATP-gamma-S >2-methylthio-ATP = ADP.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: P2RY2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR