Anti-RRM2B Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Lieferant: Bioss
BS-4181R-A350
BOSSBS-4181R-A350EA 893 EUR
BOSSBS-4181R-A350
Anti-RRM2B Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
This gene encodes the small subunit of a p53-inducible ribonucleotide reductase. This heterotetrameric enzyme catalyzes the conversion of ribonucleoside diphosphates to deoxyribonucleoside diphosphates. The product of this reaction is necessary for DNA synthesis. Mutations in this gene have been associated with autosomal recessive mitochondrial DNA depletion syndrome, autosomal dominant progressive external ophthalmoplegia-5, and mitochondrial neurogastrointestinal encephalopathy. Alternatively spliced transcript variants have been described.[provided by RefSeq, Feb 2010].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RRM2B
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat