Anti-p53R2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-4181R-A750
BOSSBS-4181R-A750EA 890 EUR
BOSSBS-4181R-A750
Anti-p53R2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
This gene encodes the small subunit of a p53-inducible ribonucleotide reductase. This heterotetrameric enzyme catalyses the conversion of ribonucleoside diphosphates to deoxyribonucleoside diphosphates. The product of this reaction is necessary for DNA synthesis. Mutations in this gene have been associated with autosomal recessive mitochondrial DNA depletion syndrome, autosomal dominant progressive external ophthalmoplegia-5, and mitochondrial neurogastrointestinal encephalopathy. Alternatively spliced transcript variants have been described.

Type: Primary
Antigen: p53R2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 131-230/351
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR