Anti-RRM2B Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-4181R-CY5.5
BOSSBS-4181R-CY5.5EA 900 EUR
BOSSBS-4181R-CY5.5
Anti-RRM2B Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
This gene encodes the small subunit of a p53-inducible ribonucleotide reductase. This heterotetrameric enzyme catalyzes the conversion of ribonucleoside diphosphates to deoxyribonucleoside diphosphates. The product of this reaction is necessary for DNA synthesis. Mutations in this gene have been associated with autosomal recessive mitochondrial DNA depletion syndrome, autosomal dominant progressive external ophthalmoplegia-5, and mitochondrial neurogastrointestinal encephalopathy. Alternatively spliced transcript variants have been described.[provided by RefSeq, Feb 2010].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RRM2B
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR