Anti-PTGER2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-4196R-A488
BOSSBS-4196R-A488EA 901 EUR
BOSSBS-4196R-A488
Anti-PTGER2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Receptor for prostaglandin E2 (PGE2). The activity of this receptor is mediated by G(s) proteins that stimulate adenylate cyclase. The subsequent raise in intracellular cAMP is responsible for the relaxing effect of this receptor on smooth muscle.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Prostaglandin E Receptor EP2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR