Anti-Prostaglandin E Receptor EP2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-4196R-A750
BOSSBS-4196R-A750EA 899 EUR
BOSSBS-4196R-A750
Anti-Prostaglandin E Receptor EP2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Receptor for prostaglandin E2 (PGE2). The activity of this receptor is mediated by G(s) proteins that stimulate adenylate cyclase. The subsequent raise in intracellular cAMP is responsible for the relaxing effect of this receptor on smooth muscle.

Type: Primary
Antigen: Prostaglandin E Receptor EP2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 271-358/358
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Sheep
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR