Anti-PTGER2 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-4196R-CY3
BOSSBS-4196R-CY3EA 901 EUR
BOSSBS-4196R-CY3
Anti-PTGER2 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Receptor for prostaglandin E2 (PGE2). The activity of this receptor is mediated by G(s) proteins that stimulate adenylate cyclase. The subsequent raise in intracellular cAMP is responsible for the relaxing effect of this receptor on smooth muscle.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Prostaglandin E Receptor EP2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR