Anti-PTGER2 Rabbit Polyclonal Antibody (Cy7®)
Lieferant: Bioss
BS-4196R-CY7
BOSSBS-4196R-CY7EA 901 EUR
BOSSBS-4196R-CY7
Anti-PTGER2 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Receptor for prostaglandin E2 (PGE2). The activity of this receptor is mediated by G(s) proteins that stimulate adenylate cyclase. The subsequent raise in intracellular cAMP is responsible for the relaxing effect of this receptor on smooth muscle.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Prostaglandin E Receptor EP2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat