Anti-HIF1AN Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-4201R-A350
BOSSBS-4201R-A350EA 893 EUR
BOSSBS-4201R-A350
Anti-HIF1AN Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
HIF1AN (Hypoxia inducible factor 1 alpha inhibitor) is a corepressor that interacts with hypoxia inducible factor 1 (HIF1) alpha and the von Hippel Lindau tumor suppressor protein to mediate repression of HIF1 transcriptional activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HIF1AN
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR