Anti-HIF1AN Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-4201R-HRP
BOSSBS-4201R-HRPEA 893 EUR
BOSSBS-4201R-HRP
Anti-HIF1AN Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
HIF1AN (Hypoxia inducible factor 1 alpha inhibitor) is a corepressor that interacts with hypoxia inducible factor 1 (HIF1) alpha and the von Hippel Lindau tumor suppressor protein to mediate repression of HIF1 transcriptional activity.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: HIF1AN
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR