Anti-Sphingomyelin Synthase 1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-4216R-A680
BOSSBS-4216R-A680EA 890 EUR
BOSSBS-4216R-A680
Anti-Sphingomyelin Synthase 1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Sphingomyelin synthases synthesize the sphingolipid, sphingomyelin, through transfer of the phosphatidyl head group, phosphatidylcholine, on to the primary hydroxyl of ceramide. The reaction is bidirectional depending on the respective levels of the sphingolipid and ceramide. Golgi apparatus SMS1 directly and specifically Recognises the choline head group on the substrate, requiring two fatty chains on the choline-P donor molecule in order to be recognized efficiently as a substrate. Major form in macrophages. Required for cell growth in certain cell types such as HeLa cells. Suppresses BAX-mediated apoptosis and also prevents cell death in response to stimuli such as hydrogen peroxide, osmotic stress, elevated temperature and exogenously supplied sphingolipids. May protect against cell death by reversing the stress-inducible increase in levels of proapoptotic ceramide.
Type: Primary
Antigen: Sphingomyelin Synthase 1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 331-413/419
Host: Rabbit
Isotype: IgG
Reactivity: Mouse