Anti-SGMS1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-4216R-CY3
BOSSBS-4216R-CY3EA 900 EUR
BOSSBS-4216R-CY3
Anti-SGMS1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Sphingomyelin synthases synthesize the sphingolipid, sphingomyelin, through transfer of the phosphatidyl head group, phosphatidylcholine, on to the primary hydroxyl of ceramide. The reaction is bidirectional depending on the respective levels of the sphingolipid and ceramide. Golgi apparatus SMS1 directly and specifically recognizes the choline head group on the substrate, requiring two fatty chains on the choline-P donor molecule in order to be recognized efficiently as a substrate. Major form in macrophages. Required for cell growth in certain cell types such as HeLa cells. Suppresses BAX-mediated apoptosis and also prevents cell death in response to stimuli such as hydrogen peroxide, osmotic stress, elevated temperature and exogenously supplied sphingolipids. May protect against cell death by reversing the stress-inducible increase in levels of proapoptotic ceramide.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SGMS1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR