Anti-UGT2B4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-4225R-A555
BOSSBS-4225R-A555EA 893 EUR
BOSSBS-4225R-A555
Anti-UGT2B4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
The UDP-Glucuronosyltransferases (UGT) comprise a family of enzymes that detoxify and enhance the urinary excretion of a wide variety of xenobiotic and endogenous substrates by transferring glucuronic acid to sulfhydryl, hydroxyl, aromatic amino, or carboxylic acid groups. They have been subdivided into two families, UGT1 and UGT2, based on the evolutionary divergence of their genes. The enzymes of the UGT1A family play an important role in the metabolism of dietary constituents, phenols, and therapeutic drugs, and also the glucuronidation of bilirubin and iodothyronines. The enzymes of the UGT2B family are involved in the metabolism of bile acids, phenol derivatives, catecholestrogens and steroids. Although it is widely recognized that the liver is the major site of glucuronidation, it is now clear that UGT enzymes are also found in extra-hepatic tissues.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: UGT2B4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR