Anti-UGT2B4 Rabbit Polyclonal Antibody (Cy7®)
Lieferant: Bioss
BS-4225R-CY7
BOSSBS-4225R-CY7EA 900 EUR
BOSSBS-4225R-CY7
Anti-UGT2B4 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
The UDP-Glucuronosyltransferases (UGT) comprise a family of enzymes that detoxify and enhance the urinary excretion of a wide variety of xenobiotic and endogenous substrates by transferring glucuronic acid to sulfhydryl, hydroxyl, aromatic amino, or carboxylic acid groups. They have been subdivided into two families, UGT1 and UGT2, based on the evolutionary divergence of their genes. The enzymes of the UGT1A family play an important role in the metabolism of dietary constituents, phenols, and therapeutic drugs, and also the glucuronidation of bilirubin and iodothyronines. The enzymes of theMore Product Information