Anti-ABAT Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-4234R-A647
BOSSBS-4234R-A647EA 893 EUR
BOSSBS-4234R-A647
Anti-ABAT Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Catalyzes the conversion of gamma-aminobutyrate and L-beta-aminoisobutyrate to succinate semialdehyde and methylmalonate semialdehyde, respectively. Can also convert delta-aminovalerate and beta-alanine.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ABAT
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR