Anti-ABAT Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-4234R-A680
BOSSBS-4234R-A680EA 890 EUR
BOSSBS-4234R-A680
Anti-ABAT Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Catalyses the conversion of gamma-aminobutyrate and L-beta-aminoisobutyrate to succinate semialdehyde and methylmalonate semialdehyde, respectively. Can also convert delta-aminovalerate and beta-alanine.

Type: Primary
Antigen: ABAT
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 151-250/500
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR