Anti-ABAT Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-4234R-CY5
BOSSBS-4234R-CY5EA 900 EUR
BOSSBS-4234R-CY5
Anti-ABAT Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Catalyzes the conversion of gamma-aminobutyrate and L-beta-aminoisobutyrate to succinate semialdehyde and methylmalonate semialdehyde, respectively. Can also convert delta-aminovalerate and beta-alanine.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ABAT
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat