Anti-ADORA1 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-4235R-CY7
BOSSBS-4235R-CY7EA 900 EUR
BOSSBS-4235R-CY7
Anti-ADORA1 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Receptor for adenosine. The activity of this receptor is mediated by G proteins which inhibit adenylyl cyclase.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADORA1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR