Anti-BMPR2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-4237R-A555
BOSSBS-4237R-A555EA 893 EUR
BOSSBS-4237R-A555
Anti-BMPR2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
On ligand binding, forms a receptor complex consisting of two type II and two type I transmembrane serine/threonine kinases. Type II receptors phosphorylate and activate type I receptors which autophosphorylate, then bind and activate SMAD transcriptional regulators. Binds to BMP-7, BMP-2 and, less efficiently, BMP-4. Binding is weak but enhanced by the presence of type I receptors for BMPs.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BMPR2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR