Anti-BMPR2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-4237R-A750
BOSSBS-4237R-A750EA 890 EUR
BOSSBS-4237R-A750
Anti-BMPR2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
On ligand binding, forms a receptor complex consisting of two type II and two type I transmembrane serine/threonine kinases. Type II receptors phosphorylate and activate type I receptors which autophosphorylate, then bind and activate SMAD transcriptional regulators. Binds to BMP-7, BMP-2 and, less efficiently, BMP-4. Binding is weak but enhanced by the presence of type I receptors for BMPs.
Type: Primary
Antigen: BMPR2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 921-1038/1038
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse