Anti-HTR3B Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-4289R-CY5.5
BOSSBS-4289R-CY5.5EA 900 EUR
BOSSBS-4289R-CY5.5
Anti-HTR3B Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
This is one of the several different receptors for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor is a ligand-gated ion channel, which when activated causes fast, depolarizing responses. It is a cation-specific, but otherwise relatively nonselective, ion channel.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HTR3B
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR